accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 244218-51-7

CH3OONHNNH2CH3HCl

CAS RN 244218-51-7

IUPAC Name: N-(4-amino-2-methylquinolin-6-yl)-2-[(4-ethylphenoxy)methyl]benzamide hydrochloride
Synonyms: N-(4-amino-2-methylquinolin-6-yl)-2-(4-ethylphenoxymethyl)benzamide hydrochloride
Molecular Weight (g/mol): 447.96
Molecular Formula: C26H26ClN3O2
InChi Key: NQLIYKXNAXKMBL-UHFFFAOYSA-N
SMILES: Cl.CCC1=CC=C(OCC2=CC=CC=C2C(=O)NC2=CC=C3N=C(C)C=C(N)C3=C2)C=C1
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results