accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 2432-14-6

CH3BrOHBr

CAS RN 2432-14-6

IUPAC Name: 2,6-dibromo-4-methylphenol
Synonyms: p-cresol, 2,6-dibromo-
Molecular Weight (g/mol): 265.93
Molecular Formula: C7H6Br2O
InChi Key: FIGPGTJKHFAYRK-UHFFFAOYSA-N
SMILES: CC1=CC(Br)=C(O)C(Br)=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)