accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 240800-45-7

FNFFF

CAS RN 240800-45-7

IUPAC Name: 5-fluoro-2-(trifluoromethyl)benzonitrile
Synonyms: 5-fluoro-2-(trifluoromethyl)benzonitrile
Molecular Weight (g/mol): 189.11
Molecular Formula: C8H3F4N
InChi Key: SQAZPEKSXOYPDX-UHFFFAOYSA-N
SMILES: FC1=CC=C(C(=C1)C#N)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results