accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 2407-11-6

ONONClS

CAS RN 2407-11-6

IUPAC Name: 2-chloro-6-nitro-1,3-benzothiazole
Synonyms: 2-chloro-6-nitro-1,3-benzothiazole
Molecular Weight (g/mol): 214.62
Molecular Formula: C7H3ClN2O2S
InChi Key: KUCSJGBXJBQHNI-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C2N=C(Cl)SC2=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results