CAS RN 2345-17-7
CAS RN 2345-17-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Irisolidone, TRC
CAS: 2345-17-7 Molecular Formula: C17H14O6 Molecular Weight (g/mol): 314.29 IUPAC Name: 5,7-dihydroxy-6-methoxy-3-(4-methoxyphenyl)chromen-4-one SMILES: COc1ccc(cc1)C2=COc3cc(O)c(OC)c(O)c3C2=O