accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 230295-09-7

H2NFFF

CAS RN 230295-09-7

IUPAC Name: 1-(2,3,6-trifluorophenyl)methanamine
Synonyms: 1-(2,3,6-trifluorophenyl)methanamine
Molecular Weight (g/mol): 161.13
Molecular Formula: C7H6F3N
InChi Key: ITWONDJIBWNPOK-UHFFFAOYSA-N
SMILES: NCC1=C(F)C=CC(F)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results