CAS RN 229027-89-8
CAS RN 229027-89-8
IUPAC Name:
(2-bromo-4-fluorophenyl)methanol
Synonyms:
(2-bromo-4-fluorophenyl)methanol
Molecular Weight (g/mol):
205.03
Molecular Formula:
C7H6BrFO
InChi Key:
XGAMLBPEVCLQEJ-UHFFFAOYSA-N
SMILES:
OCC1=C(Br)C=C(F)C=C1
1
–
1
of
1
results
1
2-Bromo-4-fluorobenzyl alcohol, 98%, Thermo Scientific™
CAS: 229027-89-8 Molecular Formula: C7H6BrFO Molecular Weight (g/mol): 205.03 MDL Number: MFCD00672925 InChI Key: XGAMLBPEVCLQEJ-UHFFFAOYSA-N PubChem CID: 2773347 IUPAC Name: (2-bromo-4-fluorophenyl)methanol SMILES: OCC1=C(Br)C=C(F)C=C1