accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3COHNH2

CAS RN 221670-72-0

IUPAC Name: 4-(1-aminoethyl)phenol
Synonyms: 1-(p-hydroxyphenyl)ethylamine
Molecular Weight (g/mol): 137.18
Molecular Formula: C8H11NO
InChi Key: CDQPLIAKRDYOCB-UHFFFAOYNA-N
SMILES: CC(N)C1=CC=C(O)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results