accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

ClClClBr

CAS RN 21928-51-8

IUPAC Name: 5-bromo-1,2,3-trichlorobenzene
Synonyms: 5-bromo-1,2,3-trichlorobenzene
Molecular Weight (g/mol): 260.34
Molecular Formula: C6H2BrCl3
InChi Key: VZUMVBQMJFFYRM-UHFFFAOYSA-N
SMILES: ClC1=CC(Br)=CC(Cl)=C1Cl
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results