Learn More
5-Bromo-1,2,3-trichlorobenzene, TRC
Product Code. 30336253
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30336253 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30336253 Supplier Toronto Research Chemicals Supplier No. TRCB688538250MG
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 5-Bromo-1,2,3-trichlorobenzene
CAS 21928-51-8
Molecular Formula C6H2BrCl3
Quantity 250 mg
Synonym 3,4,5-Trichlorobromobenzene, Bromo-3,4,5-trichlorobenzene
InChI Formula InChI=1S/C6H2BrCl3/c7-3-1-4(8)6(10)5(9)2-3/h1-2H
SMILES ClC(C(Cl)=CC(Br)=C1)=C1Cl
IUPAC Name 5-bromo-1,2,3-trichlorobenzene
Molecular Weight (g/mol) 260.34
Formula Weight 260.34