accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 21887-64-9

CH3CH3CH3OOHNH2NOHO

CAS RN 21887-64-9

IUPAC Name: 5-amino-2-{[(tert-butoxy)carbonyl]amino}pentanoic acid
Synonyms: 5-amino-2-[(tert-butoxycarbonyl)amino]pentanoic acid
Molecular Weight (g/mol): 232.28
Molecular Formula: C10H20N2O4
InChi Key: AMPVNPYPOOQUJF-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC(CCCN)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)