accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

FOHOFOH

CAS RN 214917-68-7

IUPAC Name: 2,6-difluoro-4-hydroxybenzoic acid
Synonyms: 2,6-difluoro-4-hydroxybenzoic acid
Molecular Weight (g/mol): 174.1
Molecular Formula: C7H4F2O3
InChi Key: NFIQGYBXSJQLSR-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(F)C=C(O)C=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results