CAS RN 214766-78-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Degarelix, MedChemExpress
MedChemExpress Degarelix is a competitive and reversible gonadotropin-releasing hormone receptor (GnRHR/LHRHR) antagonist. Degarelix can be used for prostate cancer research.