Learn More
Degarelix, MedChemExpress
Description
Degarelix is a competitive and reversible gonadotropin-releasing hormone receptor (GnRHR/LHRHR) antagonist. Degarelix can be used for prostate cancer research.
Specifications
Specifications
| Chemical Name or Material | Degarelix |
| Physical Form | Powder |
| CAS | 214766-78-6 |
| Color | White |
| Quantity | 50 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C82H103ClN18O16 |
| For Use With (Application) | Cancer-programmed cell death |
| Solubility Information | DMSO : 10 mg/mL (6.13 mM; Need ultrasonic) ∣H2O : 5 mg/mL (3.06 mM; ultrasonic and warming and heat to 60°C) |
| SMILES | C[C@H](C(N)=O)NC([C@H]1N(C([C@H](CCCCNC(C)C)NC([C@H](CC(C)C)NC([C@@H](CC2=CC=C(NC(N)=O)C=C2)NC([C@H](CC3=CC=C(NC([C@H](CC(N4)=O)NC4=O)=O)C=C3)NC([C@H](CO)NC([C@@H](CC5=CC=CN=C5)NC([C@@H](CC6=CC=C(Cl)C=C6)NC([C@@H](CC7=CC=C8C=CC=CC8=C7)NC(C)=O)=O)=O)=O)=O)=O)=O)=O)=O)CCC1)=O |
| Show More |