CAS RN 2140-65-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
N1-Methylguanosine, TRC
CAS: 2140-65-0 Molecular Formula: C11H15N5O5 Molecular Weight (g/mol): 297.27 Synonym: 1-Methylguanosine IUPAC Name: 2-amino-9-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1-methylpurin-6-one SMILES: CN1C(=O)C2=C(N=C1N)N(C=N2)[C@H]3[C@@H]([C@@H]([C@H](O3)CO)O)O
2
1-Methylguanosine, MedChemExpress
MedChemExpress 1-Methylguanosine is a methylated nucleoside originating from RNA degradation. 1-Methylguanosine is a tumour marker.