Learn More
1-Methylguanosine, MedChemExpress
Description
1-Methylguanosine is a methylated nucleoside originating from RNA degradation. 1-Methylguanosine is a tumour marker.
Specifications
Specifications
| Chemical Name or Material | 1-Methylguanosine |
| CAS | 2140-65-0 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C11H15N5O5 |
| Quantity | 5 mg |
| Solubility Information | DMSO : 125 mg/mL (420.49 mM; Need ultrasonic) |
| SMILES | OC[C@@H]1[C@H]([C@H]([C@H](N2C=NC3=C2N=C(N)N(C)C3=O)O1)O)O |
| Molecular Weight (g/mol) | 297.27 |
| Formula Weight | 297.27 |
| Show More |