accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 21331-80-6

BrH3CNCH3P

CAS RN 21331-80-6

IUPAC Name: [2-(dimethylamino)ethyl]triphenylphosphanium bromide
Synonyms: [2-(dimethylamino)ethyl]triphenylphosphanium bromide
Molecular Weight (g/mol): 414.33
Molecular Formula: C22H25BrNP
InChi Key: YIIHEMGEQQWSMA-UHFFFAOYSA-M
SMILES: [Br-].CN(C)CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results