CAS RN 210344-95-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Z-DEVD-FMK, MedChemExpress
MedChemExpress Z-DEVD-FMK is a specific and irreversible caspase-3 inhibitor with an IC50 of 18 μM.