Learn More
Z-DEVD-FMK, MedChemExpress
Product Code. 16477907
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16477907 5 mg 5mg
16451259 1 mg 1mg
16425188 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16477907 Supplier Medchem Express Supplier No. HY12466
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Z-DEVD-FMK is a specific and irreversible caspase-3 inhibitor with an IC50 of 18 μM.
Z-DEVD-FMK is a specific and irreversible caspase-3 inhibitor with an IC50 of 18 μM.
Specifications
Chemical Name or Material Z-DEVD-FMK
CAS 210344-95-9
Color White
Purity Grade Notes Research
Molecular Formula C30H41FN4O12
Quantity 5 mg
Solubility Information DMSO : ≥ 33.33 mg/mL (49.85 mM)
SMILES O=C(N[C@@H](C(C)C)C(N[C@H](C(CF)=O)CC(OC)=O)=O)[C@H](CCC(OC)=O)NC([C@H](CC(OC)=O)NC(OCC1=CC=CC=C1)=O)=O
Molecular Weight (g/mol) 668.66
Formula Weight 668.66
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death