accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 2103-57-3

H3COOOH3COCH3

CAS RN 2103-57-3

IUPAC Name: 2,3,4-trimethoxybenzaldehyde
Synonyms: 2,3,4-trimethoxybenzaldehyde
Molecular Weight (g/mol): 196.2
Molecular Formula: C10H12O4
InChi Key: UCTUXUGXIFRVGX-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=O)C(OC)=C1OC
Molecular Weight (g/mol) 
  • (6)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (1)
  • (6)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)