accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3OCH3H3CN

CAS RN 20662-84-4

IUPAC Name: trimethyl-1,3-oxazole
Synonyms: trimethyl-1,3-oxazole
Molecular Weight (g/mol): 111.14
Molecular Formula: C6H9NO
InChi Key: ZRLDBDZSLLGDOX-UHFFFAOYSA-N
SMILES: CC1=NC(C)=C(C)O1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results