accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 20624-92-4

CH3OOHClOH

CAS RN 20624-92-4

IUPAC Name: 2-chloro-4-(hydroxymethyl)-6-methoxyphenol
Synonyms: 2-chloro-4-(hydroxymethyl)-6-methoxyphenol
Molecular Weight (g/mol): 188.61
Molecular Formula: C8H9ClO3
InChi Key: VQEZBFOBPUKDGM-UHFFFAOYSA-N
SMILES: COC1=C(O)C(Cl)=CC(CO)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results