accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

HOOFFFFFFFFFFFFFFFFFFFFF

CAS RN 2058-94-8

IUPAC Name: henicosafluoroundecanoic acid
Synonyms: henicosafluoroundecanoic acid
Molecular Weight (g/mol): 564.09
Molecular Formula: C11HF21O2
InChi Key: SIDINRCMMRKXGQ-UHFFFAOYSA-N
SMILES: OC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results