accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 20481-17-8

IUPAC Name: 5-tert-butylbenzene-1,2,3-triol
Synonyms: 5-tert-butylbenzene-1,2,3-triol
Molecular Weight (g/mol): 182.22
Molecular Formula: C10H14O3
InChi Key: HCNISNCKPIVZDX-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC(O)=C(O)C(O)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results