accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 202865-70-1

FOHOBr(E/Z)

CAS RN 202865-70-1

IUPAC Name: 3-(2-bromo-5-fluorophenyl)prop-2-enoic acid
Synonyms: 3-(2-bromo-5-fluorophenyl)prop-2-enoic acid
Molecular Weight (g/mol): 245.05
Molecular Formula: C9H6BrFO2
InChi Key: AWWFJLNOEZAVHZ-UHFFFAOYSA-N
SMILES: OC(=O)C=CC1=C(Br)C=CC(F)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results