accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 202865-66-5

FOHBr

CAS RN 202865-66-5

IUPAC Name: (2-bromo-5-fluorophenyl)methanol
Synonyms: (2-bromo-5-fluorophenyl)methanol
Molecular Weight (g/mol): 205.03
Molecular Formula: C7H6BrFO
InChi Key: HXGZPMHPSBJMKB-UHFFFAOYSA-N
SMILES: OCC1=C(Br)C=CC(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results