accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 20262-76-4

CH3NCH3NCH3H3CSOOOSOOOOHNa

CAS RN 20262-76-4

IUPAC Name: sodium 4-{[4-(diethylamino)phenyl][4-(diethyliminiumyl)cyclohexa-2,5-dien-1-ylidene]methyl}-6-hydroxybenzene-1,3-disulfonate
Synonyms: sodium 4-{[4-(diethylamino)phenyl][4-(diethyliminio)cyclohexa-2,5-dien-1-ylidene]methyl}-6-hydroxybenzene-1,3-disulfonate
Molecular Weight (g/mol): 582.66
Molecular Formula: C27H31N2NaO7S2
InChi Key: PMLFOMWMYRKZRF-UHFFFAOYSA-M
SMILES: [Na+].CCN(CC)C1=CC=C(C=C1)C(=C1C=CC(C=C1)=[N+](CC)CC)C1=CC(O)=C(C=C1S([O-])(=O)=O)S([O-])(=O)=O
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Color 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results