Learn More
RS-127445 hydrochloride, MedChemExpress
Product Code. 16449087
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Product Code. 16449087
Brand: Medchem Express HY15419
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress RS-127445 hydrochloride is a selective, high affinity, orally bioavailable 5-HT2B receptor antagonist with a pKi of 9.5. RS-127445 hydrochloride shows 1000 fold selectivity for this receptor as compared to numerous other receptor and ion channel binding sites.
RS-127445 hydrochloride is a selective, high affinity, orally bioavailable 5-HT2B receptor antagonist with a pKi of 9.5. RS-127445 hydrochloride shows 1000 fold selectivity for this receptor as compared to numerous other receptor and ion channel binding sites.
Specifications
Chemical Name or Material RS-127445 hydrochloride
CAS 199864-86-3
Color White
Purity Grade Notes Research
Molecular Formula C17H17ClFN3
Quantity 5 mg
Solubility Information DMSO : ≥ 31 mg/mL (97.55 mM) ∣H2O : 2.27 mg/mL (7.14 mM; ultrasonic and warming and heat to 60°C)
SMILES NC1=NC(C(C)C)=CC(C2=C3C=CC=CC3=C(F)C=C2)=N1.[H]Cl
Molecular Weight (g/mol) 317.79
Formula Weight 317.79
Percent Purity 99.58%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation