accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 19808-30-1

BrNHNO

CAS RN 19808-30-1

IUPAC Name: 5-bromo-3,4-dihydropyrimidin-4-one
Synonyms: 5-bromo-3H-pyrimidin-4-one
Molecular Weight (g/mol): 174.99
Molecular Formula: C4H3BrN2O
InChi Key: CLJIGLPOBRSBNU-UHFFFAOYSA-N
SMILES: BrC1=CN=CNC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results