accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 197507-59-8

IUPAC Name: 1,6-naphthyridine-2-carboxylate
Synonyms: 1,6-naphthyridine-2-carboxylate
Molecular Weight (g/mol): 173.15
Molecular Formula: C9H5N2O2
InChi Key: OZZMWXQJCJUCEJ-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC=C2C=NC=CC2=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results