accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 19597-69-4

LiNNN

CAS RN 19597-69-4

IUPAC Name: lithium(1+) azide
Synonyms: lithium azide
Molecular Weight (g/mol): 48.96
Molecular Formula: LiN3
InChi Key: GUWHRJQTTVADPB-UHFFFAOYSA-N
SMILES: [Li+].[N-]=[N+]=[N-]
Grade 
  • (2)
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)