accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

PtClClClClClClNaNaH2OH2OH2OH2OH2OH2O

CAS RN 19583-77-8

IUPAC Name: platinum(4+) disodium hexahydrate hexachloride
Synonyms: platinum(4+) disodium hexahydrate hexachloride
Molecular Weight (g/mol): 561.85
Molecular Formula: Cl6H12Na2O6Pt
InChi Key: ZBFQOIBWJITQRI-UHFFFAOYSA-H
SMILES: O.O.O.O.O.O.[Na+].[Na+].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Pt+4]
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (4)
  • (3)
  • (4)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)