accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 19212-42-1

CH3OCH3ON

CAS RN 19212-42-1

IUPAC Name: 1-(5-methyl-3-phenyl-1,2-oxazol-4-yl)ethan-1-one
Synonyms: 1-(5-methyl-3-phenyl-1,2-oxazol-4-yl)ethanone
Molecular Weight (g/mol): 201.23
Molecular Formula: C12H11NO2
InChi Key: LWPHDEBMXBDBBO-UHFFFAOYSA-N
SMILES: CC(=O)C1=C(C)ON=C1C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results