accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 191157-01-4

OOHNO

CAS RN 191157-01-4

IUPAC Name: benzyl 6-oxo-1,6-dihydropyridine-3-carboxylate
Synonyms: benzyl 6-oxo-1H-pyridine-3-carboxylate
Molecular Weight (g/mol): 229.24
Molecular Formula: C13H11NO3
InChi Key: HGBIRINXNOIQIR-UHFFFAOYSA-N
SMILES: O=C(OCC1=CC=CC=C1)C1=CNC(=O)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results