accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

ClNCS

CAS RN 18967-44-7

IUPAC Name: 1-chloro-2-(isothiocyanatomethyl)benzene
Synonyms: 1-chloro-2-(isothiocyanatomethyl)benzene
Molecular Weight (g/mol): 183.65
Molecular Formula: C8H6ClNS
InChi Key: RMVDNJDSLXQPAV-UHFFFAOYSA-N
SMILES: ClC1=CC=CC=C1CN=C=S

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results