accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 186590-18-1

FFHOCl

CAS RN 186590-18-1

IUPAC Name: 6-chloro-2,3-difluorophenol
Synonyms: 6-chloro-2,3-difluorophenol
Molecular Weight (g/mol): 164.54
Molecular Formula: C6H3ClF2O
InChi Key: LFTQGYACSLFIRK-UHFFFAOYSA-N
SMILES: OC1=C(Cl)C=CC(F)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results