accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1805-32-9

HOClCl

CAS RN 1805-32-9

IUPAC Name: (3,4-dichlorophenyl)methanol
Synonyms: 3,4-dichlorobenzyl alcohol
Molecular Weight (g/mol): 177.02
Molecular Formula: C7H6Cl2O
InChi Key: FVJIUQSKXOYFKG-UHFFFAOYSA-N
SMILES: OCC1=CC=C(Cl)C(Cl)=C1
Molecular Weight (g/mol) 
  • (3)
  • (5)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results