accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 180200-66-2

H2OH2OH2OH3CONHNH3CFONOHOH3CONHNH3CFONOHO

CAS RN 180200-66-2

IUPAC Name: bis(1-cyclopropyl-6-fluoro-8-methoxy-7-(3-methylpiperazin-1-yl)-4-oxo-1,4-dihydroquinoline-3-carboxylic acid) trihydrate
Synonyms: bis(gatifloxacin) trihydrate
Molecular Weight (g/mol): 804.85
Molecular Formula: C38H50F2N6O11
InChi Key: RMJMZKDEVNTXHE-UHFFFAOYNA-N
SMILES: O.O.O.COC1=C2N(C=C(C(O)=O)C(=O)C2=CC(F)=C1N1CCNC(C)C1)C1CC1.COC1=C2N(C=C(C(O)=O)C(=O)C2=CC(F)=C1N1CCNC(C)C1)C1CC1
Molecular Weight (g/mol) 
  • (3)
  • (1)
Color 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
Type 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)