accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3CNCH3PClONCH3CH3

CAS RN 1794-24-7

IUPAC Name: bis(diethylamino)phosphinoyl chloride
Synonyms: didiethylaminophosphinoyl chloride
Molecular Weight (g/mol): 226.68
Molecular Formula: C8H20ClN2OP
InChi Key: HKKBJRUSFKTHSU-UHFFFAOYSA-N
SMILES: CCN(CC)P(Cl)(=O)N(CC)CC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)