accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

NOOHH2NHClHCl

CAS RN 178933-04-5

IUPAC Name: 2-amino-3-(pyridin-4-yl)propanoic acid dihydrochloride
Synonyms: 2-amino-3-(pyridin-4-yl)propanoic acid dihydrochloride
Molecular Weight (g/mol): 239.1
Molecular Formula: C8H12Cl2N2O2
InChi Key: NAMMTAFKUFJMSD-UHFFFAOYNA-N
SMILES: Cl.Cl.NC(CC1=CC=NC=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results