Learn More
D-Alanine-3,3,3-d3, CDN
Product Code. 30309712
Change view
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30309712 0.25 g 0.25g
30309672 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30309712 Supplier CDN Supplier No. CDND25600.25G
Please to purchase this item. Need a web account? Register with us today!
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material D-Alanine-3,3,3-d3
CAS 177614-69-6
Molecular Formula C3 D3 H4 N O2
Quantity 0.25 g
Synonym D-Alanine-3,3,3-d3 (9 CI, ACI)
InChI Formula InChI=1 S/C3H7NO2/c1-2(4)3(5)6/h2H,4H2,1H3,(H,5,6)/t2-/m1/s1/i1D3
SMILES [2 H]C([2 H])([2 H])[C@@H](N)C(=O)O
IUPAC Name (2 R)-2-amino-3,3,3-trideuteriopropanoic acid
Molecular Weight (g/mol) 92.1117
Formula Weight 92.0665 g/mol
Percent Purity 99 atom % D, min 98% Chemical Purity