accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 17577-28-5

ClCH3OOP

CAS RN 17577-28-5

IUPAC Name: (2-ethoxy-2-oxoethyl)triphenylphosphanium chloride
Synonyms: (2-ethoxy-2-oxoethyl)triphenylphosphanium chloride
Molecular Weight (g/mol): 384.84
Molecular Formula: C22H22ClO2P
InChi Key: DJGHVEPNEJKZBF-UHFFFAOYSA-M
SMILES: [Cl-].CCOC(=O)C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results