accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 17537-64-3

CH3ONH3CO

CAS RN 17537-64-3

IUPAC Name: 1-(1-acetyl-1H-indol-3-yl)ethan-1-one
Synonyms: 1-(1-acetylindol-3-yl)ethanone
Molecular Weight (g/mol): 201.23
Molecular Formula: C12H11NO2
InChi Key: STUZJORZRZCLRI-UHFFFAOYSA-N
SMILES: CC(=O)N1C=C(C(C)=O)C2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results