accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 17481-06-0

H3COHN(S)FOO

CAS RN 17481-06-0

IUPAC Name: (2S)-2-acetamido-3-(4-fluorophenyl)propanoate
Synonyms: (2S)-2-acetamido-3-(4-fluorophenyl)propanoate
Molecular Weight (g/mol): 224.21
Molecular Formula: C11H11FNO3
InChi Key: NRLBRFQYMSTLJK-JTQLQIEISA-M
SMILES: CC(=O)N[C@@H](CC1=CC=C(F)C=C1)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results