missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ N-Acetyl-DL-4-fluorophenylalanine, 98%
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Melting Point | 150°C to 153°C |
| CAS | 17481-06-0 |
| Molecular Formula | C11H12FNO3 |
| MDL Number | MFCD00017983 |
| Beilstein | 3206105 |
| Synonym | n-acetyl-4-fluoro-dl-phenylalanine, 2-acetamido-3-4-fluorophenyl propanoic acid, acetyl-4-fluoro-dl-phenylalanine, n-acetyl-dl-p-fluorophenylalanine, n-acetyl-dl-4-fluorophenylalanine, 2-acetylamino-3-4-fluorophenyl propanoic acid, n-acetyl-p-fluorophenylalanine, dl-phenylalanine, n-acetyl-4-fluoro, ac-dl-phe 4-f-oh, pubchem13348 |
| InChI Key | NRLBRFQYMSTLJK-UHFFFAOYSA-N |
| SMILES | CC(=O)NC(CC1=CC=C(C=C1)F)C(=O)O |
| IUPAC Name | 2-acetamido-3-(4-fluorophenyl)propanoic acid |
| Molecular Weight (g/mol) | 225.219 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction