accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

OONHNH2F

CAS RN 1737-62-8

IUPAC Name: 2-(4-fluorophenoxy)acetohydrazide
Synonyms: 2-(4-fluorophenoxy)acetohydrazide
Molecular Weight (g/mol): 184.17
Molecular Formula: C8H9FN2O2
InChi Key: SEIHQPCBPNHCBN-UHFFFAOYSA-N
SMILES: NNC(=O)COC1=CC=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results