accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3SiH3CCH3ONFOSiCH3H3CCH3N

CAS RN 17242-85-2

IUPAC Name: 5-fluoro-2,4-bis[(trimethylsilyl)oxy]pyrimidine
Synonyms: 5-fluoro-2,4-bis[(trimethylsilyl)oxy]pyrimidine
Molecular Weight (g/mol): 274.44
Molecular Formula: C10H19FN2O2Si2
InChi Key: MPVQFVDWJDULDL-UHFFFAOYSA-N
SMILES: C[Si](C)(C)OC1=NC=C(F)C(O[Si](C)(C)C)=N1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results