accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 17096-07-0

H3CCH2OOSiOSiCH3CH3H3COSiH3CCH3CH3OSiCH3H3CH3C

CAS RN 17096-07-0

IUPAC Name: 3-{2,2,6,6-tetramethyl-4-[(trimethylsilyl)oxy]-3,5-dioxa-2,4,6-trisilaheptan-4-yl}propyl 2-methylprop-2-enoate
Synonyms: 3-{2,2,6,6-tetramethyl-4-[(trimethylsilyl)oxy]-3,5-dioxa-2,4,6-trisilaheptan-4-yl}propyl 2-methylprop-2-enoate
Molecular Weight (g/mol): 422.82
Molecular Formula: C16H38O5Si4
InChi Key: BESKSSIEODQWBP-UHFFFAOYSA-N
SMILES: CC(=C)C(=O)OCCC[Si](O[Si](C)(C)C)(O[Si](C)(C)C)O[Si](C)(C)C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results