CAS RN 16941-12-1
CAS RN 16941-12-1
IUPAC Name:
dihydrogen hexachloroplatinumdiuide
Synonyms:
chloroplatinic acid
Molecular Weight (g/mol):
415.85
Molecular Formula:
Cl6H8Pt
InChi Key:
QGXPJOUUIMLZTA-UHFFFAOYSA-P
SMILES:
[H+].[H+].Cl.Cl.Cl.Cl.Cl.Cl.[Pt--]
1
–
3
of
3
results
1
Dihydrogen hexachloroplatinate(IV) solution, Pt 20% (cont. Pt)
CAS: 16941-12-1 MDL Number: MFCD00011330
2
Dihydrogen hexachloroplatinate(IV) solution, Pt 20% (cont. Pt)
CAS: 16941-12-1 | H{2}Cl{6}Pt