accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

NSOOCl

CAS RN 16851-83-5

IUPAC Name: 1-(4-chlorobenzenesulfonyl)-1H-pyrrole
Synonyms: 1-(4-chlorobenzenesulfonyl)pyrrole
Molecular Weight (g/mol): 241.69
Molecular Formula: C10H8ClNO2S
InChi Key: ACNNPPGRQRFTSO-UHFFFAOYSA-N
SMILES: ClC1=CC=C(C=C1)S(=O)(=O)N1C=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results